Sterile sigma wipes,PROSAT®
Leverandør: CONTEC
PSC20001-BPR
115-2774EA 25225 NOK
115-2774
Sterile sigma wipes,PROSAT®
Papirservietter Renromsservietter Våtservietter, renrom
Cellulose/polyester nonwoven wipes presaturated with 70% IPA and 30% DI water.
  • Suitable for Grade A/B or ISO Class 5-8 environments
  • Validated sterile per gamma irradiation
  • Hydroentangled cellulose/polyester fabric saturated with 70% USP grade IPA and 30% DI water
  • Nonwoven material is low in particles and fibres, making an excellent general purpose wipe
  • Highly sorbent with good wet strength
  • Presaturated wipe reduces VOC emissions and solvent use
  • Resealable pouch preserves cleanliness and solvent saturation levels
These PROSAT® Sterile wipes areMore Product Information