Sigma wipes, PROSAT®
Leverandør: CONTEC
PSC20002-BPR
115-2781EA 20825 NOK
115-2781
Sigma wipes, PROSAT®
Papirservietter Renromsservietter Våtservietter, renrom
Cellulose/polyester nonwoven wipes presaturated with 70% IPA and 30% DI water
  • Suitable for ISO Class 5-8 environments
  • Nonwoven material is low in particles and fibres, making an excellent general purpose wipe
  • Highly sorbent with good wet strength
  • Presaturated to reduce solvent usage and VOC emissions
PROSAT® Sigma wipes consist of cellulose/polyester hydroentangled wipes saturated with a solution of 70% USP grade isopropanol and 30% deionized water. They are ideal for use in life science applications.
Cellulose/polyester wipes are a cost-effectiveMore Product Information