Cleanroom wipes, pre-saturated, SATWipes®
Leverandør: CONTEC
SWNW0013-BPR
115-2828EA 4975 NOK
115-2828
Cleanroom wipes, pre-saturated, SATWipes®
Papirservietter Renromsservietter Våtservietter, renrom
These wipes consist of 50% polyester and 50% rayon (non woven) that is hydraulically entangled creating a soft, durable, and clean wipe. The saturation of 70% alcohol and 30% deionized water provides a safe and mild cleaning solution.
  • Strong, sorbent and low linting
  • Eliminate in-house blending
  • Reduces potential for cross-contamination
These wipes offer a convenient and controlled solution to wet wiping applications. Ideal for use in animal lab facilities.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904