Interior and surface cleaners, SURE®
Leverandør: Diversey
100892011
115-2961EA 745 NOK
115-2961
Interior and surface cleaners, SURE®
Rengjøringsløsninger Detergenter og overflatevasking
Concentrated all-purpose detergent for cleaning of hard surfaces, such as floors, walls, doors, ceilings and general equipment. Can also be used in food premises.
  • Suitable for cleaning of all water resistant hard surfaces, including aluminium and other soft metals
  • Effective in all water conditions
  • No fragrance or dye
  • Contains plant-based ingredients from renewable sources decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for user
  • Concentrated formulation gives excellent economy of use
  • Fast acting, fast drying
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Sustainable Materials,Safer Human and Environmental Health,Sustainable Packaging