Dual-wire metal wristband, MagSnap 360™
Leverandør: DESCO
19887
115-3036EA 475 NOK
115-3036
Dual-wire metal wristband, MagSnap 360™
Bånd mot elektrostatisk utladning Håndleddbånd mot elektrostatisk utladning
MagSnap 360™ dual-wire metal wristband creates a superior electrical connection to eliminate intermittent failures.
  • Magnetic connection between wristband and coil cord
  • Medical grade series stainless steel backplate and links
  • Date coded
Magnetic connection between wristband and coil cord creates a superior electrical connection, and eliminates intermittent failures in traditional snap connections using springs, extends the lifespan of the wrist strap, and maintains a 1 to 5 pound perpendicular breakaway force to prevent accidental disconnects.More Product Information