Dissipative floor finish, 10 L box, Statguard®
Leverandør: DESCO
220521
115-3038EA 1875 NOK
115-3038
Dissipative floor finish, 10 L box, Statguard®
Rengjøringsløsninger Detergenter og overflatevasking
Statguard® Static Dissipative Cross-Linked Floor Finish is used to dissipate static charges as well as prevent triboelectric (static charge generation) charges while providing a clear, high gloss finish that resists wear. It meets EN 61340-5-1 ESD Floors Table 1 RG less than or equal to 1x10E9 ohms.
  • Non-tribocharging
  • Static dissipative
  • Converts a non-ESD floor to an ESD floor
  • Ensures worker safety and mitigates user’s liability exposure
  • Easy to apply; improves productivity
  • Cross-linked polymer; produces high-gloss, durable finish
  • Low humidity formulation