Elastic adjustable cam-lock wrist band
Leverandør: DESCO
229600
115-3039EA 75 NOK
115-3039
Elastic adjustable cam-lock wrist band
Bånd mot elektrostatisk utladning Håndleddbånd mot elektrostatisk utladning
  • Versatile; metal back or plastic back
  • Metal helps contact to dry skin - insert plastic cover if there are metal-on-skin issues
  • Adjustable band to suit wrist circumference of 130 to 180 mm
  • 10 mm stud connector only