Dual-wire wrist strap parking station, MagSnap 360™
Leverandør: DESCO
231431
115-3046EA 200 NOK
115-3046
Dual-wire wrist strap parking station, MagSnap 360™
Bånd mot elektrostatisk utladning Jordingsledning
This provides a storage terminal for the wrist cord when the workstation is unoccupied.
  • Mounts to any surface conveniently located nearby the operator
  • Spares operator jacks from the mechanical wear and tear of constant insertion and removal of the wrist cord
  • Monitors display a PASS condition for the operator when the wrist cord is parked
Gross weight: 0,01 kg.