Interconnect mat cord, SCS
Leverandør: DESCO
2389
115-3047EA 295 NOK
115-3047
Interconnect mat cord, SCS
Bånd mot elektrostatisk utladning Jordingsledning
Daisy-chains two worksurface mats for the monitoring of two workstations with the 724 workstation monitor.
  • Reduces inadvertent snagging that may disconnect the cord from the worksurface mat
  • No resistor
This monitor/table mat interconnect cord is a straight cord with male stud snap fasteners at each end, and no resistors.
Sertifiseringer: Meets ANSI/ESD S20.20 and Grounding standard ANSI/ESD S6.1.