Surface and mat cleaner, Reztore®
Leverandør: DESCO
10446
115-3048EA 210 NOK
115-3048
Surface and mat cleaner, Reztore®
Rengjøringsløsninger Detergenter og overflatevasking
Reztore® Surface and mat cleaners removes dust, grease, grime, fingerprints, solder flux and other contaminants from ESD mats and other surfaces.
  • One quart trigger spray bottle
  • New alcohol-free formula
  • Non-hazardous, non-flammable green chemistry formula
  • No silicone or insulative residues
No special handling requirements for shipping and storage; may be used in normal workplace environments. Cleaners with silicone leave an insulative layer preventing conductive or dissipative mats from functioning properly.
Sertifiseringer: Lead-free RoHS compliant.