Non-marking foot grounders, 3-layers
Leverandør: DESCO
17252
115-3049EA 145 NOK
115-3049
Non-marking foot grounders, 3-layers
Bånd mot elektrostatisk utladning Hælbånd mot elektrostatisk utladning
These grounders will not mark shoes or floors - reducing maintenance costs of touching up on floors. Size adjustable material expands to fit most shoe sizes comfortably and flex during walking.
  • Neoprene rubber with no exposed carbon
  • Hook and loop fastening system
  • 76,2 cm long, eight conductive strand nylon polyester grounding tab
  • One megohm series resistor
  • Date coded
  • Made in the United States of America
Additional conductive material provides reliable connection with operator’s sweat-layer. To protect operator from accidental contact with equipmentMore Product Information