Single wire disposable wrist strap, SCS
Leverandør: DESCO
2209
115-3055EA 22 NOK
115-3055
Single wire disposable wrist strap, SCS
Bånd mot elektrostatisk utladning Håndleddbånd mot elektrostatisk utladning
Wrist strap (2209) is designed for one-time use. The wrist strap is ideally suited for shipment along with ESD sensitive items [ESDS], providing the means to ground personnel opening ESDS packaging and installing contents.
  • Disposable non-woven fabric wrist strap provides cost-effective
  • Hypo-allergenic adhesive provides 360 degree contact with skin
  • Minimizes skin-to-band resistance
  • Conductive adhesive on the copper foil adheres to any convenient electrical ground
Use for visitors and one-time use applications. Suited for shipment along with componentsMore Product Information