Elastic adjustable wrist straps, Jewel®
Leverandør: DESCO
60001
115-3068EA 140 NOK
115-3068
Elastic adjustable wrist straps, Jewel®
Bånd mot elektrostatisk utladning Håndleddbånd mot elektrostatisk utladning
High visibility bright colour allows supervisor to see that worker is wearing wrist strap.
  • Adjustable
  • Locking clasp, band locked so it will not slip
  • Extra wide, soft elastic band for increased wearer comfort
  • Non-shedding, non-sloughing band material, safe for microelectronics
  • Nickel-plated brass wristband stud machined to exacting tolerances and precise profile
  • Silver impregnated polypropylene knitted on inside for superior conductivity
Replacement elastic bands available; and easy to replace without tools. Unique adjustment design eliminates exposedMore Product Information