BenchGuard®/BenchGuard® Extra, Sterilin® Bench Protectors
Leverandør: STERILIN
BG50E BG50 BG60E
115-9321EA 5675 NOK
115-9321 115-9305 115-9320
BenchGuard®/BenchGuard® Extra, Sterilin® Bench Protectors
Benkepapir Benkepapir
Highly absorbent paper for protecting benches and surfaces against liquid spills.
  • One side plastic coated to prevent soak through
  • Available in either roll or sheet forms
  • BenchGuard Extra®: double absorbency for more demanding applications
Absorption of 400 ml/m² for BenchGuard® and 800 ml/m² for BenchGuard® Extra.
Leveringsinformasjon: Supplied in easy to use dispenser packs.