2,2'-Bipyridyl 99+%

Leverandør: Thermo Fisher Scientific
Danger

117500100 117500250 117501000 117505000
117500100.EA 845 NOK
117500100. 117500250. 117501000. 117505000.
2,2'-Bipyridyl 99+%
2,2'-Bipyridyl
Formel: C₁₀H₈N₂
Kokepunkt: 272…273 °C (760 mmHg)
Smeltepunkt: 69…72 °C
Tetthet: 1,326 g/cm³ (-150 °C)
Flash punkt: 121 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006212
CAS-nummer: 366-18-7
EINECS: 206-674-4
FN: 2811
ADR: 6.1,III
Merck Indeks: 13,03382
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e86bbc29edb6c5d09c058b8efcb6b9eb7f4d070c&vid=9e3a03e37045643109ae0765802e1997d30604ae&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 69 to 72 °C
GC ≥99.0 %
Suitability (for Fe determination) Passes test
Barium (Ba) ≤0.001 %
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤5 ppm
Manganese (Mn) ≤0.001 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR