Plaster dispenser, Salvequick
Leverandør: Cederroth / Orkla Care AB
490700
118-0159EA 460 NOK
118-0159
Plaster dispenser, Salvequick
Førstehjelpsutstyr Bandasjer
This dispenser for adhesive bandages (plasters) keeps the plasters clean and dry until time of use. When a plaster is needed it can easily be pulled out and applied on the wound with one hand.
  • Dispenser also functions as an anti-theft system
  • Each plaster is sterile and individually wrapped
  • Refills are locked, which prevents waste
  • Quick and easy to refill with a special key
  • Transparent lid protects against dust
The green-coloured plastic in Cederroth Stations and Dispensers are from January 2022 made of >85% recycled plastics. Old refrigerator partsMore Product Information