Eyewash station and solution, Maxi
Leverandør: Plum Safety
4707 4708
118-0238EA 365 NOK
118-0238 118-0239
Eyewash station and solution, Maxi
Førstehjelpsutstyr Øyeskyllestasjon
This wall station is equipped with two 1000 ml eyewash bottles, wall brackets, pictogram and mirror. The single bottle contains sterile sodium chloride solution (0,9%). It is equipped with an ergonomic eye cup and a dust cover.
  • Station is particularly suitable for use in workplaces where a longer rinsing time is required
  • Bottle can be used for refilling the eyewash station.
Total rinsing time of the station: 20 minutes
W×D×H of the station: 300×132×240 mm
Shelf life: 3 years