Eye and wound cleansing spray
Leverandør: Cederroth / Orkla Care AB
726000
118-0247EA 310 NOK
118-0247
Eye and wound cleansing spray
Førstehjelpsutstyr Førstehjelpssett, enkeltdeler
This cleansing spray is intended for rinsing and cleaning out dust and dirt from eyes and wounds. The portable spray bottle is filled with 150 ml physiological (0,9%) unbuffered sodium chloride solution.
  • Solution is preservative-free and remains sterile until last drop
  • Spray is suitable for use on both eyes and wounds
  • Actuator is protected by a plastic cap and a tamper seal
  • Nitrogen propellant is not in contact with the solution