Eyewash station
Leverandør: Cederroth / Orkla Care AB
721500
118-0277EA 3550 NOK
118-0277
Eyewash station
Førstehjelpsutstyr Øyeskyllestasjon
Wall-mountable eyewash station with two 500 ml bottles which open automatically when they are twisted out of their holders. The solution works to neutralise eye pH, i.e. it has a neutralising effect on any alkali or acid splashes, giving a quicker result than a normal sodium chloride solution.
  • Contains Salvequick plaster dispenser (incl. 40 sterile fabric and 45 sterile plastic plasters), eyewash instructions and a key
  • Features extra space for other accessories, e.g. Bloodstopper dressings, protective glasses or gloves
  • Transparent door protects the