Eyewash station, QuickSafe® Chemical Industry
Leverandør: Plum Safety
5171
118-0379EA 1925 NOK
118-0379
Eyewash station, QuickSafe® Chemical Industry
Førstehjelpsutstyr Øyeskyllestasjon
This wall-mountable station is made of impact resistant plastic and equipped with a transparent cover. The station is specifically designed for use in the chemical industry, laboratories and in other companies where employees are in close contact with acids and alkalis.
  • Contains Duo eyewash to rinse both eyes at once
  • Cabinet is only slightly larger than an A3 sheet of paper
  • Comes with instructions and fixtures for wall mounting
Contents:
1×500 ml Plum Eye Wash Duo
1×200 ml pH Neutral
1×500 ml pH Neutral Duo
5×QuickRinse eyewash ampoules (20 ml)
1More Product Information