Fall protection harnesses, Miller H-Design® Duraflex™ 2 pts harness Auto 2 loops
Leverandør: HONEYWELL SAFETY
1032872 1032871 1032873
118-0440EA 2775 NOK
118-0440 113-8649 113-8650
Fall protection harnesses, Miller H-Design® Duraflex™ 2 pts harness Auto 2 loops
Fallvern Fallsele
Miller H-Design® Duraflex™ 2 points harness with two webbing loops at the front and automatic buckles on chest and leg straps. The Miller H-Design® harnesses are dedicated to construction and general industry sectors, providing more comfort, freedom of movement and safety to workers.
  • Straps are made from Duraflex Stretch and black webbing (PES with PTFE treatment)
  • Straps are water repellent and anti-stain treated
  • Sternal anchorage point with long webbing loops
  • Steel/zinc plated silver finished back anchorage D-ring and mating buckles
  • Aluminium automatic