Lockout hasp, non conductive
Leverandør: Brady
236916
118-0533EA 245 NOK
118-0533
Lockout hasp, non conductive
Sikkerhetslåser
This non conductive nylon hasp can accommodate up to six locks, withstands up to 450 kg, yet only weighs 30 g.
  • Lockout shackle has holes with a minimum Ø of 7 mm
  • Made of orange nylon
Max. shackle Ø: 9,00 mm
Min. shackle clearance: 20,00 mm