Øyevaskeflaske, pH-nøytral
Leverandør: Plum Safety
4752 4755
118-1707EA 240 NOK
118-1707 118-0479
Øyevaskeflaske, pH-nøytral
Førstehjelpsutstyr Øyeskyllestasjon
Steril fosfatbufferløsning (4,9 %) for rask og effektiv nøytralisering av syrer og baser.
  • Handy sized bottle, well suited for use in first aid kits and emergency bags
  • Bottle with ergonomic eye cup, dust cap and clear instructions
  • Can be used as refill for Plum wall-mounted eyewash station
  • No maintenance and cleaning of the bottles required
  • Bottles in single carton: Carton protects the bottles from external contaminations and can be opened quickly and easily at the perforation
Rinsing time: ~2 minutes per 200 ml
Shelf life: 3 years
DIN EN 15154-4More Product Information