Ruthenium(IV) oxide hydrate (≥54% Ru)

Leverandør: Thermo Fisher Scientific
Warning

11803.06 11803.03 11803.14
11803.06EA 6950 NOK
11803.06 11803.03 11803.14
Ruthenium(IV) oxide hydrate (≥54% Ru)
Ruthenium(IV) oxide hydrate

CAS No.: 32740-79-7 Molecular Formula: O2Ru.xH2O Formula Weight: 133.07(anhy) Physical Form: Powder Appearance: Black Odour: Odourless UN No.: 1479 MDL No.: MFCD00149846


Oxidation

Formel: O₂Ru·H₂O
Smeltepunkt: 1200 °C (subl.)
Tetthet: 6,97 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149846
CAS-nummer: 32740-79-7
EINECS: 234-840-6
ADR: ,

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR