Benedict's reagent Sugar_urine_qual, Supelco®

Leverandør: Merck
Warning


MQ100

11945-250ML
11945-250MLEA 730 NOK
11945-250ML
Benedict's reagent Sugar_urine_qual, Supelco®
Benedict's Reagent

Reagent used for the detection of reducing sugars and other reducing substances; detection limit: 0,01% glucose in water.


Benedict′s reagent may be used during TLC analysis of mesocotyl methanol extracts during a study to evaluate sorghum resistance associated with accumulation of undetected toxic host metabolites. It may also be also be used to synthesize an SEM image of the cubic shaped Cu₂O with smooth surface.


Benedict′s reagent is comprised of copper sulphate, sodium carbonate which functions as a mild alkali, and sodium citrate. It is quite stable. On boiling this reagent with glucose, it gives a white precipitate of cuprous thiocyanate. It is primarily used for the estimation of glucose in urine

Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00146068
CAS-nummer: 63126-89-6
FN: 3082
ADR: 9 III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR