PIG® Oil-Only absorbent socks
Leverandør: NEW PIG
SKM210
121-0128EA 1625 NOK
121-0128
PIG® Oil-Only absorbent socks
Materialer for håndtering av søl
PIG® Oil-Only Absorbent Socks absorbs and retains oils and oil-based liquids - including lubricants and fuels - without absorbing a drop of water.
  • Outlasts other outdoor socks with the most UV resistant construction
  • Moldable socks hug corners and surround machine bases to absorb machine leaks, contain puddles and keep workers safe
  • Cellulose filler is hydrophobic for containing and absorbing oil-based spills in a variety of environments
  • Polypropylene skin resists tearing; reduces dust and holds in liquid, even when saturated
  • Ideal for use on land, but also