Spill kits, ADR
Leverandør: Brady
813955 813866 813864 813867
121-0154EA 1600 NOK
121-0154 121-0153 121-0152 121-0193
Spill kits, ADR
Materialer for håndtering av søl
ADR spill kits are eco-friendly and portable and come in a durable see-through bag with shoulder strap. The small ADR chemical kit is designed for small spills of chemical and hazardous liquid. The large ADR maintenance kit is designed for small spills of industrial oil, water, solvents and coolants. The Re-Form ADR kit S is designed for small spills of industrial liquids except for highly aggressive acids and caustics.
  • Quick and easy access to the sorbents
  • Durable see-through bag makes it easy to check the spill response supplies