VWR®, Spill Kit Chemical, 16 L
Leverandør: VWR Collection
121-0194EA 1100 NOK
121-0194
VWR®, Spill Kit Chemical, 16 L
Materialer for håndtering av søl
This portable, economy spill kit comes in a high visibility, water-resistant, yellow PVC bag with handles. Designed for small spills of chemicals and hazardous liquids. Ideal for small storage areas, thanks to its compact size.
  • Fast spill response
  • Yellow colour for high visibility
  • Water-resistant, lightweight bag keeps sorbents dry
  • Can fit behind or under a seat in a vehicle
All you need for an emergency spill response in a compact and high vis carry bag. Ideal for workers on the go, like field workers, or for carriers and truck drivers whoMore Product Information