VWR®, Spill Kit, Acid Neutralising
Leverandør: VWR Collection
121-0196EA 2275 NOK
121-0196
VWR®, Spill Kit, Acid Neutralising
Materialer for håndtering av søl
This speciality spill kit contains sorbents, neutralising agent, PPE and accessories so that you are prepared in an emergency. Contained in a handy tub, providing 17 litres of absorption capacity.
  • Suitable for most acid spills
  • Lightweight and easy to move for a fast response
  • Highly absorbing pads to wipe down equipment and remove spills
  • SOC’s to prevent spills from spreading on the work floor
  • Neutralising polymer to neutralise acid
  • Gloves and glasses to protect employees
  • Waste disposal bag to contain all used equipment
Position spill kits around yourMore Product Information