VWR®, Spill Kit, Base Neutralising
Leverandør: VWR Collection
121-0197EA 2375 NOK
121-0197
VWR®, Spill Kit, Base Neutralising
Materialer for håndtering av søl
This speciality spill kit contains sorbents, neutralising agent, PPE and accessories so that you are prepared in an emergency. Contained in a handy tub, providing 17 litres of absorption capacity.
  • Use to neutralise highly basic substances before clean-up
  • Lightweight and easy to move for a fast response
  • Highly absorbing pads to wipe down equipment and remove spills
  • SOC’s to prevent spills from spreading on the work floor
  • Neutralising polymer to neutralise bases
  • Gloves and glasses to protect employees
  • Waste disposal bag to contain all used equipment