Kaliumcyanid ≥96.0% ACS

Leverandør: Thermo Fisher Scientific
Danger

12136.18 12136.30 12136.A1
12136.18EA 365 NOK
12136.18 12136.30 12136.A1
Kaliumcyanid ≥96.0% ACS
Potassium cyanide
Formel: KCN
Kokepunkt: 1625 °C
Smeltepunkt: 634 °C
Tetthet: 1,52
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011397
CAS-nummer: 151-50-8
EINECS: 205-792-3
FN: 1680
ADR: 6.1,I
Merck Indeks: 14,7626

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR