Nikkel(II)nitrat heksahydrat Ni ≥ 19,8%, tech.

Leverandør: Thermo Fisher Scientific
Danger

12222.36 12222.A3
12222.36EA 755 NOK
12222.36 12222.A3
Nikkel(II)nitrat heksahydrat Ni ≥ 19,8%, tech.
Nikkel(II)nitrat heksahydrat

CAS No.: 13478-00-7 Molecular Formula: N2NiO6.6H2O Formula Weight: 290.80 (182.70anhy) Physical Form: Flake Density: 2.05 Sensitivity: Hygroscopic Appearance: Green Odour: Odourless Melting Point: 56.7° Boiling Point: 137° Solubility: Soluble in water, alcohol Merck Reference: 14,6513 UN No.: 2725 MDL No.: MFCD00149805


Nickel plating, manufacturing of ceramic Colours

Formel: Ni(NO₃)₂·6H₂O
Kokepunkt: 137 °C
Tetthet: 2,05
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149805
CAS-nummer: 13478-00-7
EINECS: 236-068-5
FN: 2725
ADR: 5.1,III
Merck Indeks: 14,6513

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR