Benzosyre ≥99.5%, certified reference material for titrimetry, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

12353-25G-F
12353-25G-FEA 3075 NOK
12353-25G-F
Benzosyre ≥99.5%, certified reference material for titrimetry, Supelco®
Benzosyre

  • certified by BAM

Formel: C₇H₆O₂
MW: 122.12 g/mol
Kokepunkt: 249
Smeltepunkt: 121…125 °C
Tetthet: 1.32 g/cm³ (20 °C)
Flash punkt: 121
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002398
CAS-nummer: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Indeks: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR