Lead selenide ≥99%

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: Lead(II) selenide

12840.18
12840.18EA 0 NOK
12840.18
Lead selenide ≥99%
Lead selenide
Formel: PbSe
MW: 286.16 g/mol
Smeltepunkt: 79.5…80 °C
Tetthet: 8.1 g/cm³ (20 °C)
MDL nummer: MFCD00016273
CAS-nummer: 12069-00-0
EINECS: 235-109-4
FN: 3283
ADR: 6.1 III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR