Adjustable handles for brooms and squeegees
Leverandør: PERFEX
53200-100
129-0127EA 960 NOK
129-0127
Adjustable handles for brooms and squeegees
Koster
These polymer handles are developed to meet the ergonomic and hygienic needs of controlled environments. They are widely accepted as a superior alternative to fixed length handles based on traditional materials, such as steel, aluminium and wood.
  • Comfortable and easy to use
  • Durable and lightweight
  • Handles will not corrode, oxidise, or rot
  • Compatible with gamma and ETO
  • Autoclavable up to 121 °C for 30 minutes
These handles are impervious to water, grease, detergents, sanitisers, and solvents. Grasp the upper handle and twist centre grip to adjust theMore Product Information