Steriliseringsposer
Leverandør: Brand
759705 759710
129-0131EA 455 NOK
129-0131 129-9812
Steriliseringsposer
Poser Avfallsposer
Bags: Disposal bag, PP/PA.
Holders:
Stativene er laget av epoksyresinbelagt ståltråd. Gummiføtter. Ikke-autoklaverbar.
Epoxy-coated steel wire, with rubber feet. Non-autoclavable.
NB: risk of injury. Never put sharp objects such as needles into plastic bags. Bags should stay open during sterilisation.