AlphaMop™ isolator cleaning tool
Leverandør: Texwipe
TX7101
129-0273EA 3350 NOK
129-0273
AlphaMop™ isolator cleaning tool
Mopper
The ITW Texwipe® Mini AlphaMop™ isolator cleaning tool is designed to clean isolators, biosafety cabinets, glove boxes and laminar flow hoods. The mop has a low, flat profile and rounded corners. It is made from Celcon® plastic and is totally autoclavable. The swivel joint allows the user to reach difficult-to-access areas. The replaceable foam pad ensures that the mop cover conforms to the surfaces being cleaned. 129-0273 includes two handles, 12 and 18'' (30,4 and 45,7 cm), and is easily used with one hand.
  • Strong and lightweight
  • Easily used in hard-to-access areas; mop covers are