VWR®, Sterile Cleanroom Double-Pouch Twist-Seal Bags
Leverandør: VWR Collection
129-9889EA 7375 NOK
129-9889 129-9890 129-0665 129-9000 129-9003 129-9001 129-9002 129-9004 129-9005 129-9006 129-9007 129-9008 129-9009 129-9010 129-9011 129-9012 129-9013 129-9014 129-9015 129-9016 129-9017 129-9018
VWR®, Sterile Cleanroom Double-Pouch Twist-Seal Bags
Poser Prøveposer
PE bags, transparent. These sterile sampling bags provide a secure, contaminant-free pliant container that ensures dependable analysis results. Economical and efficient way to collect, contain and carry samples. With these bags, the user can bring sterile sampling bags into a cleanroom without any worries about contamination. The bags are double-pouched so the user can remove the outer bag before entering the cleanroom, which minimises the number of bags exposed.
  • The bags are sterilised inside and out by gamma irradiation
  • Sterility certificate in every box