Spatula
Leverandør: BURKLE
5383-1000
129-1158EA 5750 NOK
129-1158
Spatula
Spatler
This spatula is made of premium stainless steel, high-gloss polished and completely welded, thus completely free of undercuts.
  • Very sturdy
  • Easy to clean
  • Autoclavable, can be flame-sterilised
  • Can pierce solid substances or sacks and BigBags
Ideal for taking samples of powders, granulates and pasty media, especially in food and pharmaceutical applications.