VWR®, Slider Zip Bags
Leverandør: VWR Collection
129-1198EA 2550 NOK
129-1198 129-1199 129-1200 129-1201 129-1202 129-1203
VWR®, Slider Zip Bags
Poser Oppbevaringsposer
VWR® slider zip bags, in LDPE, transparent, with blue slider. For the packing and storage of goods.
  • Can be used from −10 to +65 °C
  • Do not contain any ingredients of animal origin
Sertifiseringer: Manufactured under ISO 9001:2015 standard.
Forpakning: 50 pcs/outer bag, 10 outer bags/case.