VWR®, Autoclavable Bags
Leverandør: VWR Collection
129-1575EA 10175 NOK
129-1575
VWR®, Autoclavable Bags
Poser Avfallsposer
Sturdy, temperature resistant bags, ideal for sterilising animal cages.
  • PE, clear
  • Autoclavable up to 121 °C
  • Highly puncture resistant and impact strength