Bags, stand-up, recyclable, PE/EVOH-PE
Leverandør: Flexico
ITM080501 ITM080502 ITM080503 ITM080504 ITM080505 ITM080506 ITM080507 ITM080508
129-2303EA 5875 NOK
129-2303 129-2304 129-2305 129-2306 129-2307 129-2308 129-2309 129-2310
Bags, stand-up, recyclable, PE/EVOH-PE
Poser Prøveposer
Clear bags, zip seal, food contact.
  • Oxygen barrier and protection against humidity
  • Ideal for greasy or dry products
  • Suitable for food contact
  • Transparency and zip seal with sound and tactile feel
  • Resealable, reusable, 100% recyclable
  • Upright position for shelf display
The PE/EVOH-PE bag is a 100% recyclable mono-material complex film. The presence of a 5 µm EVOH layer provides excellent barrier protection against oxygen, carbon dioxide and retains aromas, without impacting the recyclability of the bag. The layers of polyethylene surrounding the EVOHMore Product Information