BHD Autoclavable Muff Covers, with Elastic
Leverandør: KEYSTONE CLEANROOM PRODUCTS
MFNWB04 MFNWB05 MFNWB06 MFNWB07 MFNWB08 MFNWB09 MFNWB12 MFNWB16 MFNWB18 MFNWB18SK02 MFNWB18SK02TL MFNWB05-SP MFNWB08-SP MFNWB12-SP MFNWB03
129-1760EA 25625 NOK
129-1760 129-1761 129-1762 129-1763 129-1764 129-1765 129-1766 129-1767 129-1768 129-2474 129-2475 129-3059 129-3062 129-3064 129-3058
BHD Autoclavable Muff Covers, with Elastic
Deksler
Save time and standardise activities in equipment preparation for sterilisation.
  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies conformance when wrapping to SOP's
  • Steam sterilisation indicator - autoclavable up to 135 °C
  • Tear and puncture resistant
  • Significant process improvement compared to wrapping with sheets and tape
BHD muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closures are breathable and can be used to prevent particulatesMore Product Information