VWR®, Self-wringing Mopping System
Leverandør: VWR Collection
129-2500EA 7500 NOK
129-2500 129-2502 129-2506 129-2508
VWR®, Self-wringing Mopping System
Mopper
This mopping system can be used without a wringer. It is ideal for applying disinfectants to floors, walls and ceilings. The high performance foam for cleanrooms enables excellent, lasting liquid transfer.
  • Autoclavable; resistant to gamma rays and ETO
  • Resistant to the most widely available disinfectants and detergents
  • Sponges made from laminated polyester, for liquid absorption and highly delicate disinfection
  • Replacement foam mop heads are available with stainless steel or galvanised steel clips