Humidity indicator cards
Leverandør: DESCO
4HIC100 6HIC200
129-2642EA 240 NOK
129-2642 129-2644
Humidity indicator cards
Testpapir Indikatorpapir
Humidity indicator cards (HICs) are printed with moisture sensitive spots which respond to various levels of humidity with a visible colour change from blue to pink.
  • Accuracy based on ambient temperature of 75 °F
  • Tolerance is approximately 2,5% for each 10 °F away from 75 °F
The moisture sensitive spots are made of blotting paper impregnated with cobaltous chloride. The humidity inside dry packed barrier bags can be monitored by including an HIC inside. When the bag is opened, the card can be examined for proper dryness inside the bag.