Autoclave Bags, Self-Seal, BHD
Leverandør: KEYSTONE CLEANROOM PRODUCTS
BNW0638SS2P BNW0840SS2P BNW0844SS2P BNW1240SS2P BNW1622SS2P BNW1648SS2P BNW2226SS2P BNW2626SS2P BNW2646SS2P BNW2832SS2P BNW4040SS2P BNW4446SS2P BNW4448SS2P BNW5858SS2P
129-2825EA 30325 NOK
129-2825 129-2826 129-2827 129-2828 129-2829 129-2830 129-2831 129-2796 129-2761 129-2762 129-2763 129-2764 129-2765 129-2766
Autoclave Bags, Self-Seal, BHD
Poser Steriliseringsposer
Self-sealing BHD autoclave bags can be used as either the inner, or outer bag for the steam sterilisation of pharmaceutical change parts.
  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies conformance when wrapping to SOP's
  • Autoclavable up to 135 °C
  • Tear and puncture resistant 2 ply blue BHD non woven
  • Significant process improvement compared to wrapping with sheets and tape
Standardise and simplify the way equipment is wrapped for steam sterilisation.
The use of these 2 ply self-sealing bags will also saveMore Product Information