Autoclave Bags with Steam Indicator, DuPont™ Tyvek® 1422A
Leverandør: KEYSTONE CLEANROOM PRODUCTS
BGT2A0.7505 BGT2A0.7508 BGT2A0105 BGT2A0106 BGT2A0108 BGT2A0205 BGT2A0306 BGT2A0404 BGT2A0406 BGT2A0409 BGT2A0416 BGT2A0430 BGT2A0610 BGT2A0612 BGT2A0638 BGT2A0820 BGT2A09512 BGT2A1015 BGT2A1030 BGT2A1062 BGT2A1212 BGT2A1216 BGT2A1218 BGT2A1232 BGT2A1234 BGT2A1240 BGT2A1246 BGT2A1248 BGT2A1416 BGT2A1418 BGT2A1424 BGT2A1543 BGT2A1604 BGT2A1612 BGT2A1624 BGT2A1638 BGT2A1824 BGT2A1830 BGT2A1836 BGT2A2020 BGT2A2024 BGT2A2030 BGT2A2040 BGT2A2224 BGT2A2424 BGT2A2436 BGT2A2442 BGT2A2448 BGT2A2462 BGT2A2628 BGT2A2844 BGT2A3016 BGT2A3020 BGT2A3036 BGT2A3040 BGT2A3060 BGT2A3412 BGT2A3636 BGT2A3642 BGT2A3648 BGT2A3660 BGT2A4242 BGT2A4416 BGT2A4848 BGT2A4864 BGT2A5054 BGT2A0110 BGT2A0124 BGT2A0203 BGT2A0208 BGT2A0236 BGT2A0250 BGT2A0309 BGT2A0310 BGT2A0412 BGT2A0448 BGT2A0516 BGT2A0522 BGT2A0526 BGT2A0608 BGT2A0662 BGT2A0710 BGT2A0808 BGT2A0809 BGT2A0812 BGT2A0816 BGT2A0822 BGT2A0824 BGT2A0856 BGT2A0872 BGT2A1020 BGT2A1024 BGT2A1222 BGT2A1224 BGT2A1236 BGT2A1252 BGT2A1420 BGT2A1422 BGT2A1426 BGT2A1526 BGT2A1530 BGT2A1534 BGT2A1620 BGT2A1632 BGT2A1818 BGT2A1848 BGT2A1855 BGT2A2026 BGT2A2028 BGT2A2044 BGT2A2222 BGT2A2225 BGT2A2226 BGT2A2325 BGT2A2430 BGT2A2440 BGT2A2460 BGT2A2472 BGT2A2818 BGT2A2834 BGT2A2838 BGT2A2840 BGT2A2864 BGT2A3030 BGT2A3246 BGT2A3434 BGT2A4040 BGT2A4836 BGT2A4860 BGT2A4872 BGT2A5050 BGT2A0506 BGT2A0606
129-3372EA 38750 NOK
129-3372 129-3373 129-3379 129-3381 129-3384 129-3192 129-3213 129-3223 129-3168 129-3167 129-3163 129-3160 129-3260 129-3263 129-3270 129-3290 129-3232 129-3235 129-3246 129-3252 129-3256 129-3593 129-3596 129-3616 129-3619 129-3624 129-3629 129-3547 129-3557 129-3559 129-3572 129-3664 129-3667 129-3669 129-3676 129-3697 129-3682 129-3632 129-3637 129-3651 129-3654 129-3659 129-3434 129-3445 129-3450 129-3455 129-3461 129-3462 129-3399 129-3402 129-3418 129-3421 129-3423 129-3430 129-3500 129-3501 129-3509 129-3524 129-3529 129-3536 129-3543 129-3479 129-3483 129-3489 129-3495 129-2797 129-3386 129-3388 129-3189 129-3198 129-3205 129-3206 129-3216 129-3218 129-3164 129-3171 129-3179 129-3181 129-3182 129-3186 129-3272 129-3277 129-3281 129-3282 129-3285 129-3289 129-3224 129-3226 129-3227 129-3228 129-3239 129-3243 129-3603 129-3609 129-3621 129-3554 129-3563 129-3570 129-3575 129-3586 129-3589 129-3590 129-3673 129-3681 129-3686 129-3643 129-3648 129-3657 129-3658 129-3437 129-3444 129-3447 129-3448 129-3449 129-3453 129-3458 129-3398 129-3401 129-3409 129-3414 129-3416 129-3417 129-3420 129-3427 129-3506 129-3513 129-3472 129-3488 129-3494 129-3498 129-3153 129-3173 129-3184
Autoclave Bags with Steam Indicator, DuPont™ Tyvek® 1422A
Poser Steriliseringsposer
Tyvek® 1422A autoclave bags with steam indicators can be used as either the inner, or outer bag for the steam sterilisation of pharmaceutical change parts.
  • Tear and puncture resistant
  • Designed to be heat sealed closed
  • Superior to paper based autoclave pouches and wraps
  • Fast and consistent for equipment preparation area
  • Steam indicator and hydrophobic
  • Lint-free; strong factory seals
  • Exceptional microbial barrier
Standardise and simplify the way equipment is wrapped for steam sterilisation. The use of these bags will also save valuable time in theMore Product Information