BinLiners™ and BinLiner™ Stands
Leverandør: MICRONOVA
BL2-40TRIR BL2-40TRIRB BL4-40TRIRB BL4-40TRIRBB BL4-40TRIR BL2-40TRB BL1-40TRIRBB BL4-4060BB BL4-4060IRBB BL3-2436IRBB BL4-3440PC BL6-4048PC BL2-1016IRBB BLT-12IR BL2-40TRBB BL3-8265PC
129-1090EA 11325 NOK
129-1090 129-1091 129-1093 129-1094 129-1092 129-1584 129-1595 129-1588 129-1587 129-1688 129-1689 129-1690 129-1687 129-1774 MICRBL2-40TRBB MICRBL3-8265PC 129-3991
BinLiners™ and BinLiner™ Stands
Poser Avfallsposer
LDPE Micronova BinLiners™ are ideal for the collection and disposal of waste, collection and transportation of parts or raw materials, and storage in cleanrooms and controlled environments. Thickness of 1 to 6 mm available for a variety of applications; including covering equipment, holding production waste, document storage, lining large production totes and so on.
  • Made with FDA compliant material
  • Single-seam construction
  • Gusseted base seals for a better fit in rectangular containers
  • Plastic core roll pack and flat pack options
  • Available in multiple sizes