Glyceryl Dibehenate, Reference Standard
Leverandør: USP
1295709
1295709.EA 5375 NOK
1295709.
Glyceryl Dibehenate, Reference Standard
Standarder Organiske standarder
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Sertifiseringer: US Pharmacopea Reference Standard
Bestill nå
SPESIFIKASJONER
Beskrivelse Glyceryl Dibehenate
Synonym(s) Glyceryl dibehenate
Environmentally Preferable
CAS-nr 18641-57-1
Formel C₆₉H₁₃₄O₆