[D-Tyr5]-Gonadorelin, Reference Standard

Leverandør: USP

1296712
1296712.EA 15925 NOK
1296712.
[D-Tyr5]-Gonadorelin, Reference Standard
Standarder Organiske standarder

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.


Sertifiseringer: US Pharmacopea Reference Standard

Bestill nå

SPESIFIKASJONER

Beskrivelse [D-Tyr5]-Gonadorelin
Synonym(s) 5-Oxo-L-prolyl-L-histidyl-L-tryptophyl-L-seryl-D-tyrosylglycyl-L-leucyl-L-arginyl-L-prolylglycinamide
Environmentally Preferable
CAS-nr 53634-21-2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR